Assay Details
Target Gene Details
Entrez Gene ID: | 138881 |
Gene Name: | olfactory receptor family 1 subfamily L member 8 |
Gene Aliases: |
OR9-24 |
Location: |
Chr.9:122546009-122576960 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1L8 | NM_001004454.1 | 1 | 204 | NP_001004454.1 |
| XM_017014284.1 | 3 | 740 | XP_016869773.1 | |
| XM_017014285.1 | 3 | 740 | XP_016869774.1 | |
| XM_017014286.1 | 2 | 611 | XP_016869775.1 | |
| BC140910.1 | 1 | 286 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054800 | Chr.9:122555206 - 122666496 on Build GRCh38 | Loss |
|
| nsv1039785 | Chr.9:122544123 - 122644710 on Build GRCh38 | Gain |
|
| esv2659552 | Chr.9:122540811 - 122598369 on Build GRCh38 | Deletion |
|
| nsv1052657 | Chr.9:122565623 - 122577487 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map