Assay Details
Target Gene Details
Entrez Gene ID: | 55958 |
Gene Name: | kelch like family member 9 |
Gene Aliases: |
- |
Location: |
Chr.9:21331019-21335430 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL9 | NM_018847.2 | 1 | 4070 | NP_061335.1 |
| AB037775.1 | 1 | 4009 | ||
| AK023630.1 | 1 | 2714 | BAB14623.1 | |
| AK025468.1 | 1 | 1946 | ||
| AL713669.1 | 1 | 4012 | CAD28475.1 | |
| AY172949.1 | AAO39718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map