Assay Details
Target Gene Details
Entrez Gene ID: | 10610 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 |
Gene Aliases: |
SAITL1, SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM |
Location: |
Chr.17:76565379-76586066 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC2 | NM_006456.2 | NP_006447.2 | ||
| XM_005256954.3 | XP_005257011.1 | |||
| AJ251053.1 | CAB61434.1 | |||
| AK225943.1 | ||||
| BT019972.1 | AAV38775.1 | |||
| U14550.1 | AAA52228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641290 | Chr.17:76547444 - 76586380 on Build GRCh38 | Gain |
|
| esv3641291 | Chr.17:76553052 - 76573475 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map