Assay Details
Target Gene Details
Entrez Gene ID: | 83461 |
Gene Name: | cell division cycle associated 3 |
Gene Aliases: |
GRCC8, TOME-1 |
Location: |
Chr.12:6844796-6852066 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA3 | NM_001297602.1 | NP_001284531.1 | ||
| NM_001297603.1 | NP_001284532.1 | |||
| NM_001297604.1 | NP_001284533.1 | |||
| NM_031299.5 | NP_112589.1 | |||
| NR_123727.1 | ||||
| AK092246.1 | ||||
| AK291421.1 | ||||
| AK312843.1 | ||||
| BC002551.2 | AAH02551.1 | |||
| BM911472.1 | ||||
| BX099267.1 | ||||
| CD106434.1 | ||||
| DA735127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss |
|
| nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
|
| esv3580405 | Chr.12:6826466 - 6903969 on Build GRCh38 | Gain |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map