Assay Details
Target Gene Details
Entrez Gene ID: | 9770 |
Gene Name: | Ras association domain family member 2 |
Gene Aliases: |
CENP-34, RASFADIN |
Location: |
Chr.20:4780023-4823668 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF2 | NM_014737.2 | 12 | 2168 | NP_055552.1 |
| NM_170774.1 | 11 | 2026 | NP_739580.1 | |
| XM_005260895.3 | 12 | 2234 | XP_005260952.1 | |
| XM_011529410.1 | 12 | 2168 | XP_011527712.1 | |
| XM_011529411.1 | 12 | 2240 | XP_011527713.1 | |
| XM_011529412.2 | 11 | 2089 | XP_011527714.1 | |
| XM_017028149.1 | 11 | 2063 | XP_016883638.1 | |
| XM_017028150.1 | 13 | 2371 | XP_016883639.1 | |
| XM_017028151.1 | 13 | 2338 | XP_016883640.1 | |
| XM_017028152.1 | 13 | 2415 | XP_016883641.1 | |
| XM_017028153.1 | 11 | 2116 | XP_016883642.1 | |
| AK307232.1 | ||||
| AY154470.1 | 12 | 2168 | AAN59975.1 | |
| AY154471.1 | 9 | 1942 | AAN59976.1 | |
| AY154472.1 | 11 | 2026 | AAN59977.1 | |
| BC117118.1 | 11 | 1977 | ||
| BC117120.1 | 10 | 2005 | ||
| CR627436.1 | 11 | 5375 | CAH10522.1 | |
| D79990.1 | 12 | 2168 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061866 | Chr.20:4781931 - 4813599 on Build GRCh38 | Gain |
|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
| nsv585310 | Chr.20:4586645 - 4794039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map