Assay Details
Target Gene Details
Entrez Gene ID: | 381 |
Gene Name: | ADP ribosylation factor 5 |
Gene Aliases: |
- |
Location: |
Chr.7:127588352-127591705 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARF5 | NM_001662.3 | NP_001653.1 | ||
| AF493884.1 | AAM12598.1 | |||
| AK297026.1 | ||||
| AK307297.1 | ||||
| AK308548.1 | ||||
| AK312107.1 | ||||
| BC003043.1 | AAH03043.1 | |||
| BC033104.1 | AAH33104.1 | |||
| BT007087.1 | AAP35750.1 | |||
| CN259625.1 | ||||
| M57567.1 | AAA90927.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831122 | Chr.7:127503153 - 127717831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map