Assay Details
Target Gene Details
Entrez Gene ID: | 8936 |
Gene Name: | WAS protein family member 1 |
Gene Aliases: |
SCAR1, WAVE, WAVE1 |
Location: |
Chr.6:110099817-110189874 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759462 | Chr.6:110068723 - 110183477 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map