Assay Details
Target Gene Details
Entrez Gene ID: | 79664 |
Gene Name: | interactor of little elongation complex ELL subunit 2 |
Gene Aliases: |
BRCC1, NARG2 |
Location: |
Chr.15:60419609-60479160 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICE2 | NM_001018089.2 | 16 | 5062 | NP_001018099.1 |
| NM_024611.5 | 16 | 5222 | NP_078887.2 | |
| XM_005254661.1 | 17 | 5221 | XP_005254718.1 | |
| XM_005254662.1 | 17 | 5084 | XP_005254719.1 | |
| XM_011522009.1 | 16 | 5096 | XP_011520311.1 | |
| XM_011522010.1 | 16 | 4965 | XP_011520312.1 | |
| XM_017022569.1 | 17 | 5258 | XP_016878058.1 | |
| AL833122.1 | 5 | 3203 | CAH56202.1 | |
| BC140894.1 | 16 | 5141 | ||
| CR749852.1 | 16 | 5015 | CAH18700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569609 | Chr.15:60417705 - 60502450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map