Assay Details
Target Gene Details
Entrez Gene ID: | 11280 |
Gene Name: | sodium voltage-gated channel alpha subunit 11 |
Gene Aliases: |
FEPS3, HSAN7, NAV1.9, NaN, PN5, SCN12A, SNS-2 |
Location: |
Chr.3:38845764-39051986 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN11A | NM_001287223.1 | NP_001274152.1 | ||
| NM_014139.2 | NP_054858.2 | |||
| XM_011533321.2 | XP_011531623.1 | |||
| XM_017005647.1 | XP_016861136.1 | |||
| XM_017005648.1 | XP_016861137.1 | |||
| XM_017005649.1 | XP_016861138.1 | |||
| XM_017005650.1 | XP_016861139.1 | |||
| XM_017005651.1 | XP_016861140.1 | |||
| XM_017005652.1 | XP_016861141.1 | |||
| XM_017005653.1 | XP_016861142.1 | |||
| AF109737.1 | AAF24976.1 | |||
| AF150882.1 | AAF24980.1 | |||
| AF188679.1 | AAF17480.1 | |||
| AJ417790.1 | CAD10507.1 | |||
| AY686224.1 | AAT95434.1 |
More Information
Set Membership: |
|

Genomic Map