Assay Details
Target Gene Details
Entrez Gene ID: | 3034 |
Gene Name: | histidine ammonia-lyase |
Gene Aliases: |
HIS, HSTD |
Location: |
Chr.12:95972662-95996365 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAL | NM_001258333.1 | 20 | 2539 | NP_001245262.1 |
| NM_001258334.1 | 20 | 2536 | NP_001245263.1 | |
| NM_002108.3 | 21 | 2606 | NP_002099.1 | |
| XM_011538249.2 | 12 | 1439 | XP_011536551.1 | |
| XM_017019246.1 | 12 | 1927 | XP_016874735.1 | |
| AK297004.1 | 8 | 1313 | ||
| AK298736.1 | 20 | 2515 | ||
| AK303544.1 | 20 | 2465 | ||
| D16626.1 | 21 | 2480 | BAA04047.1 |
More Information
Set Membership: |
|

Genomic Map