Assay Details
Target Gene Details
Entrez Gene ID: | 686 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.3:15601352-15653709 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTD | AK297033.1 |
Target Gene Details
Entrez Gene ID: | 26061 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
2-HPCL, HPCL, HPCL2, PHYH2 |
Location: |
Chr.3:15560704-15601852 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACL1 | NM_001284413.1 | 1 | 237 | NP_001271342.1 |
| NM_001284415.1 | 1 | 237 | NP_001271344.1 | |
| NM_001284416.1 | 1 | 237 | NP_001271345.1 | |
| NM_012260.3 | 1 | 237 | NP_036392.2 | |
| NR_104315.1 | 1 | 237 | ||
| AK301990.1 | ||||
| DB049608.1 | ||||
| DB241165.1 | 1 | 237 |
More Information
Additional Information:
For this assay, SNP(s) [rs115586700] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map