Assay Details
Target Gene Details
Entrez Gene ID: | 65018 |
Gene Name: | PTEN induced putative kinase 1 |
Gene Aliases: |
BRPK, PARK6 |
Location: |
Chr.1:20633455-20651511 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PINK1 | NM_032409.2 | 8 | 2233 | NP_115785.1 |
| AB053323.1 | 8 | 2233 | BAB55647.1 | |
| AF316873.1 | 8 | 2209 | AAK28062.1 | |
| AK075225.1 | 4 | 1232 | BAC11484.1 | |
| AK130746.1 | 5 | 2698 | ||
| BC009534.1 | 5 | 1306 | AAH09534.1 | |
| BC028215.1 | 8 | 2148 | AAH28215.1 |
Target Gene Details
Entrez Gene ID: | 100861548 |
Gene Name: | PINK1 antisense RNA |
Gene Aliases: |
NAPINK1, PINK1-AS1, PINK1AS |
Location: |
Chr.1:20642657-20652193 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PINK1-AS | NR_046507.1 | 1 | 1110 | |
| AK090466.1 | 1 | 1110 | ||
| AK172749.1 | 1 | 195 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv1003348 | Chr.1:20648970 - 20715062 on Build GRCh38 | Gain |
|
| nsv834369 | Chr.1:20609770 - 20779837 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map