Assay Details
Target Gene Details
Entrez Gene ID: | 3636 |
Gene Name: | inositol polyphosphate phosphatase like 1 |
Gene Aliases: |
OPSMD, SHIP2 |
Location: |
Chr.11:72223861-72239147 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPPL1 | NM_001567.3 | NP_001558.3 | ||
| XM_005273978.3 | XP_005274035.1 | |||
| XM_005273979.4 | XP_005274036.1 | |||
| XM_011544999.2 | XP_011543301.1 | |||
| XM_011545000.2 | XP_011543302.1 | |||
| XM_017017673.1 | XP_016873162.1 | |||
| BC140853.1 | ||||
| L36818.1 | AAA96658.1 | |||
| Y14385.1 | CAA74743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951038 | Chr.11:72226157 - 72253456 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map