Assay Details
Target Gene Details
Entrez Gene ID: | 85021 |
Gene Name: | RALBP1 associated Eps domain containing 1 |
Gene Aliases: |
RALBP1 |
Location: |
Chr.6:138904015-138988282 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPS1 | NM_001128617.2 | 19 | 3395 | NP_001122089.1 |
| NM_001286611.1 | 20 | 3476 | NP_001273540.1 | |
| NM_001286612.1 | 17 | 3203 | NP_001273541.1 | |
| NM_031922.4 | 20 | 3473 | NP_114128.3 | |
| XM_005267178.4 | 19 | 3391 | XP_005267235.1 | |
| XM_017011388.1 | 19 | 2777 | XP_016866877.1 | |
| AI879474.1 | 1 | 75 | ||
| CA424106.1 | 1 | 93 |
More Information
Set Membership: |
|

Genomic Map