Assay Details
Target Gene Details
Entrez Gene ID: | 6566 |
Gene Name: | solute carrier family 16 member 1 |
Gene Aliases: |
HHF7, MCT, MCT1, MCT1D |
Location: |
Chr.1:112911847-112956353 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A1 | NM_001166496.1 | 4 | 2011 | NP_001159968.1 |
| NM_003051.3 | 4 | 1542 | NP_003042.3 | |
| XM_011542026.2 | 4 | 1275 | XP_011540328.1 | |
| XM_011542027.2 | 4 | 1285 | XP_011540329.1 | |
| AK000641.1 | 1 | 427 | ||
| AK094064.1 | 4 | 2011 | ||
| AK296688.1 | 4 | 1325 | ||
| AK303603.1 | 4 | 1384 | ||
| AK312500.1 | 4 | 1385 | ||
| AL162079.1 | 4 | 1372 | CAB82412.1 | |
| BC026317.1 | 4 | 1385 | AAH26317.1 | |
| BC045664.1 | 5 | 2096 | AAH45664.1 | |
| L31801.1 | 3 | 1191 | AAC41707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584026 | Chr.1:112761723 - 113036481 on Build GRCh38 | Gain |
|
| nsv1004102 | Chr.1:112753183 - 113008588 on Build GRCh38 | Gain |
|
| esv24626 | Chr.1:112832815 - 113310193 on Build GRCh38 | Loss |
|
| nsv999915 | Chr.1:112817523 - 113142211 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map