Assay Details
Target Gene Details
Entrez Gene ID: | 166614 |
Gene Name: | doublecortin like kinase 2 |
Gene Aliases: |
CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2 |
Location: |
Chr.4:150078274-150258563 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCLK2 | NM_001040260.3 | 1 | 782 | NP_001035350.2 |
| NM_001040261.4 | 1 | 782 | NP_001035351.4 | |
| NR_036614.1 | 1 | 782 | ||
| XM_005262785.2 | 1 | 611 | XP_005262842.1 | |
| XM_005262786.2 | 1 | 611 | XP_005262843.1 | |
| XM_017007832.1 | 1 | 611 | XP_016863321.1 | |
| XM_017007833.1 | 1 | 611 | XP_016863322.1 | |
| AB209181.1 | 1 | 782 | BAD92418.1 | |
| DA122884.1 | 1 | 251 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830111 | Chr.4:149946205 - 150114404 on Build GRCh38 | Gain |
|
| nsv949829 | Chr.4:150078249 - 150079948 on Build GRCh38 | Deletion |
|
| esv3894052 | Chr.4:149995950 - 150249235 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map