Assay Details
Target Gene Details
Entrez Gene ID: | 4985 |
Gene Name: | opioid receptor delta 1 |
Gene Aliases: |
OPRD |
Location: |
Chr.1:28812142-28863696 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPRD1 | NM_000911.3 | 1 | 405 | NP_000902.3 |
| BF434068.1 | 1 | 437 | ||
| EF057065.1 | 1 | 163 | ||
| EU883570.1 | 1 | 163 | ||
| U07882.2 | 1 | 405 | AAA18789.2 | |
| U10504.1 | 1 | 163 | AAA83426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525310 | Chr.1:28812463 - 28818676 on Build GRCh38 | Loss |
|
| esv3585594 | Chr.1:28808979 - 28813822 on Build GRCh38 | Loss |
|
| nsv545921 | Chr.1:28781383 - 28839626 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111622802] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map