Assay Details
Target Gene Details
Entrez Gene ID: | 30815 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
SIAT7-F, SIAT7F, ST6GALNACVI |
Location: |
Chr.9:127885321-127906725 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv497n21 | Chr.9:127870863 - 127898531 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map