Assay Details
Target Gene Details
Entrez Gene ID: | 89 |
Gene Name: | actinin alpha 3 (gene/pseudogene) |
Gene Aliases: |
- |
Location: |
Chr.11:66546395-66563329 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTN3 | NM_001104.3 | NP_001095.2 | ||
| NM_001258371.2 | NP_001245300.2 | |||
| NR_047663.2 | ||||
| AK125851.1 | ||||
| AK303044.1 | ||||
| AK313556.1 | ||||
| BC096323.2 | AAH96323.1 | |||
| BC099647.3 | AAH99647.1 | |||
| BC099648.1 | AAH99648.1 | |||
| BC099649.3 | AAH99649.1 | |||
| M86407.1 | AAA51585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626696 | Chr.11:66552406 - 66562756 on Build GRCh38 | Loss |
|
| esv2744639 | Chr.11:66552336 - 66562723 on Build GRCh38 | Deletion |
|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map