Assay Details
Target Gene Details
Entrez Gene ID: | 10482 |
Gene Name: | nuclear RNA export factor 1 |
Gene Aliases: |
MEX67, TAP |
Location: |
Chr.11:62792125-62805492 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXF1 | NM_001081491.1 | 3 | 422 | NP_001074960.1 |
| NM_006362.4 | 3 | 422 | NP_006353.2 | |
| AB209915.1 | 3 | 357 | BAD93152.1 | |
| AF112880.1 | 3 | 286 | AAD39102.1 | |
| AF126246.1 | 3 | 286 | AAD20016.1 | |
| AJ132712.1 | 3 | 286 | CAA10753.1 | |
| AK304082.1 | ||||
| AK304137.1 | 3 | 401 | ||
| AK304743.1 | ||||
| AK307961.1 | 4 | 528 | ||
| AK307998.1 | 3 | 370 | ||
| BC004904.2 | 3 | 359 | AAH04904.1 | |
| BC028041.1 | 3 | 352 | AAH28041.1 | |
| BM751313.1 | 3 | 411 | ||
| U80073.1 | 2 | 106 | AAB81111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508636 | Chr.11:62779642 - 62876986 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map