Assay Details
Target Gene Details
Entrez Gene ID: | 7422 |
Gene Name: | vascular endothelial growth factor A |
Gene Aliases: |
MVCD1, VEGF, VPF |
Location: |
Chr.6:43770209-43786487 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEGFA | NM_001025366.2 | 1 | 694 | NP_001020537.2 |
| NM_001025367.2 | 1 | 694 | NP_001020538.2 | |
| NM_001025368.2 | 1 | 694 | NP_001020539.2 | |
| NM_001025369.2 | 1 | 694 | NP_001020540.2 | |
| NM_001025370.2 | 1 | 694 | NP_001020541.2 | |
| NM_001033756.2 | 1 | 694 | NP_001028928.1 | |
| NM_001171622.1 | 1 | 694 | NP_001165093.1 | |
| NM_001171623.1 | 1 | 694 | NP_001165094.1 | |
| NM_001171624.1 | 1 | 694 | NP_001165095.1 | |
| NM_001171625.1 | 1 | 694 | NP_001165096.1 | |
| NM_001171626.1 | 1 | 694 | NP_001165097.1 | |
| NM_001171627.1 | 1 | 694 | NP_001165098.1 | |
| NM_001171628.1 | 1 | 694 | NP_001165099.1 | |
| NM_001171629.1 | 1 | 694 | NP_001165100.1 | |
| NM_001171630.1 | 1 | 694 | NP_001165101.1 | |
| NM_001204384.1 | 1 | 694 | NP_001191313.1 | |
| NM_001204385.1 | 1 | 694 | NP_001191314.1 | |
| NM_001317010.1 | 1 | 694 | NP_001303939.1 | |
| NM_003376.5 | 1 | 694 | NP_003367.4 | |
| AF022375.1 | 1 | 364 | AAC63143.1 | |
| BC011177.1 | 1 | 169 | ||
| BC065522.1 | 1 | 121 | ||
| BU153227.1 | 1 | 131 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823682 | Chr.6:43770248 - 43771276 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map