Assay Details
Target Gene Details
Entrez Gene ID: | 1073 |
Gene Name: | cofilin 2 |
Gene Aliases: |
NEM7 |
Location: |
Chr.14:34710382-34714823 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFL2 | NM_001243645.1 | NP_001230574.1 | ||
| NM_021914.7 | NP_068733.1 | |||
| NM_138638.4 | NP_619579.1 | |||
| NR_028130.1 | ||||
| NR_028131.1 | ||||
| XM_011536363.2 | XP_011534665.1 | |||
| AF087867.1 | AAM10495.1 | |||
| AF134802.1 | AAD31280.1 | |||
| AF134803.1 | AAD31281.1 | |||
| AK314157.1 | ||||
| AL599958.1 | ||||
| BC011444.1 | AAH11444.1 | |||
| BC022364.1 | AAH22364.1 | |||
| BC022876.1 | AAH22876.1 | |||
| CB992605.1 | ||||
| DA298739.1 | ||||
| DA897112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041857 | Chr.14:34615683 - 34947974 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map