Assay Details
Target Gene Details
Entrez Gene ID: | 151636 |
Gene Name: | deltex E3 ubiquitin ligase 3L |
Gene Aliases: |
BBAP |
Location: |
Chr.3:122564338-122575203 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3L | NM_138287.3 | 1 | 158 | NP_612144.1 |
| AF484416.1 | 1 | 69 | AAL90859.1 | |
| AK125086.1 | 1 | 158 | ||
| AL833598.1 | 1 | 91 | ||
| AY225123.1 | 1 | 203 | AAP57517.1 | |
| AY780792.1 | 1 | 69 | AAV98362.1 | |
| BC042191.1 | 1 | 77 | AAH42191.1 | |
| BC060509.1 | 1 | 100 | AAH60509.1 |
Target Gene Details
Entrez Gene ID: | 83666 |
Gene Name: | poly(ADP-ribose) polymerase family member 9 |
Gene Aliases: |
ARTD9, BAL, BAL1, MGC:7868 |
Location: |
Chr.3:122527910-122564676 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP9 | NM_001146103.1 | 1 | 182 | NP_001139575.1 |
| NM_031458.2 | 1 | 182 | NP_113646.2 | |
| AF307338.1 | 1 | 83 | AAK07558.1 | |
| AF307339.1 | 1 | 83 | AAK07559.1 | |
| DB096326.1 | 1 | 278 |
More Information
Set Membership: |
|

Genomic Map