Assay Details
Target Gene Details
Entrez Gene ID: | 1314 |
Gene Name: | coatomer protein complex subunit alpha |
Gene Aliases: |
AILJK, HEP-COP |
Location: |
Chr.1:160288587-160343564 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPA | NM_001098398.1 | NP_001091868.1 | ||
| NM_004371.3 | NP_004362.2 | |||
| BC038447.1 | AAH38447.1 | |||
| U24105.1 | AAB70879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3210 | Chr.1:160253051 - 160298030 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map