Assay Details
Target Gene Details
Entrez Gene ID: | 23001 |
Gene Name: | WD repeat and FYVE domain containing 3 |
Gene Aliases: |
ALFY, ZFYVE25 |
Location: |
Chr.4:84669537-84966391 on Build GRCh38 |
Assay Gene Location: | Within Exon 71 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY3 | NM_014991.4 | 65 | 10422 | NP_055806.2 |
| XM_005262858.4 | 67 | 10463 | XP_005262915.1 | |
| XM_011531757.2 | 66 | 10374 | XP_011530059.1 | |
| XM_011531759.2 | 65 | 10336 | XP_011530061.1 | |
| XM_011531760.2 | 65 | 10274 | XP_011530062.1 | |
| XM_011531761.2 | 66 | 10329 | XP_011530063.1 | |
| XM_011531762.2 | 65 | 10323 | XP_011530064.1 | |
| XM_011531763.2 | 64 | 10220 | XP_011530065.1 | |
| XM_011531764.2 | 65 | 10290 | XP_011530066.1 | |
| XM_011531765.2 | 62 | 10025 | XP_011530067.1 | |
| XM_017007906.1 | 68 | 10591 | XP_016863395.1 | |
| XM_017007907.1 | 68 | 10588 | XP_016863396.1 | |
| XM_017007908.1 | 67 | 10520 | XP_016863397.1 | |
| XM_017007909.1 | 53 | 8216 | XP_016863398.1 | |
| AB023210.2 | 30 | 4105 | BAA76837.2 | |
| AF538685.1 | 64 | 10314 | AAN15137.1 | |
| AK055806.1 | BAB71020.1 | |||
| AK092394.1 | 5 | 801 | ||
| BC013377.2 | 2 | 766 | AAH13377.1 | |
| BC015214.1 | 5 | 748 | ||
| BC065502.1 | 12 | 1772 | AAH65502.1 | |
| BC119633.2 | 2 | 469 | ||
| BC148294.1 | 2 | 469 | ||
| BC148298.2 | 2 | 469 | ||
| BC150246.1 | 30 | 4126 | ||
| BX648077.1 | 22 | 2987 | ||
| BX648153.1 | 25 | 3315 |
More Information
Set Membership: |
|

Genomic Map