Assay Details
Target Gene Details
Entrez Gene ID: | 100129931 |
Gene Name: | uncharacterized LOC100129931 |
Gene Aliases: |
- |
Location: |
Chr.4:7030554-7046231 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129931 | NR_033828.1 | 4 | 2747 | |
| AK126065.1 | 4 | 2747 |
Target Gene Details
Entrez Gene ID: | 57533 |
Gene Name: | TBC1 domain family member 14 |
Gene Aliases: |
- |
Location: |
Chr.4:6909444-7033118 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D14 | NM_001113361.1 | 14 | 3876 | NP_001106832.1 |
| NM_001113363.1 | 12 | 3142 | NP_001106834.1 | |
| NM_001286805.1 | 12 | 3011 | NP_001273734.1 | |
| NM_020773.2 | 14 | 3998 | NP_065824.2 | |
| XM_005247986.2 | 13 | 3917 | XP_005248043.1 | |
| XM_006713895.2 | 14 | 4166 | XP_006713958.1 | |
| XM_011513507.2 | 15 | 4085 | XP_011511809.1 | |
| XM_011513508.1 | 15 | 3978 | XP_011511810.1 | |
| XM_011513513.2 | 13 | 3180 | XP_011511815.1 | |
| XM_017008477.1 | 13 | 3804 | XP_016863966.1 | |
| XM_017008478.1 | 13 | 3248 | XP_016863967.1 | |
| XM_017008479.1 | 11 | 3012 | XP_016863968.1 | |
| XM_017008480.1 | 15 | 3310 | XP_016863969.1 | |
| AB037743.1 | 13 | 3826 | ||
| AL833868.1 | 11 | 2863 | CAD38726.1 | |
| BC041167.1 | 14 | 3921 | AAH41167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428436 | Chr.4:6986421 - 7268174 on Build GRCh38 | Gain |
|
| esv34033 | Chr.4:6971543 - 7388894 on Build GRCh38 | Loss |
|
| nsv469847 | Chr.4:7001798 - 7166714 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map