Assay Details
Target Gene Details
Entrez Gene ID: | 79961 |
Gene Name: | DENN domain containing 2D |
Gene Aliases: |
- |
Location: |
Chr.1:111185969-111204791 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND2D | NM_001271833.1 | NP_001258762.1 | ||
| NM_024901.4 | NP_079177.2 | |||
| XM_006710921.2 | XP_006710984.2 | |||
| XM_006710922.1 | XP_006710985.1 | |||
| XM_006710923.1 | XP_006710986.1 | |||
| XM_006710924.1 | XP_006710987.1 | |||
| XM_011542187.1 | XP_011540489.1 | |||
| XM_011542189.1 | XP_011540491.1 | |||
| XM_011542190.2 | XP_011540492.1 | |||
| XM_017002386.1 | XP_016857875.1 | |||
| XM_017002387.1 | XP_016857876.1 | |||
| XM_017002388.1 | XP_016857877.1 | |||
| AK026110.1 | BAB15363.1 | |||
| AK301583.1 | ||||
| BC004557.2 | AAH04557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map