Assay Details
Target Gene Details
Entrez Gene ID: | 4103 |
Gene Name: | MAGE family member A4 |
Gene Aliases: |
CT1.4, MAGE-41, MAGE-X2, MAGE4, MAGE4A, MAGE4B |
Location: |
Chr.X:151912889-151925170 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA4 | NM_001011548.1 | 3 | 1576 | NP_001011548.1 |
| NM_001011549.1 | 3 | 1573 | NP_001011549.1 | |
| NM_001011550.1 | 3 | 1564 | NP_001011550.1 | |
| NM_002362.4 | 3 | 1579 | NP_002353.3 | |
| XM_005274677.2 | 3 | 1619 | XP_005274734.1 | |
| XM_005274678.3 | 3 | 1577 | XP_005274735.1 | |
| XM_005274679.3 | 3 | 1586 | XP_005274736.1 | |
| AY954624.1 | 2 | 1449 | AAY27261.1 | |
| BC017723.1 | 3 | 1576 | AAH17723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510866 | Chr.X:151858097 - 151930089 on Build GRCh38 | Deletion |
|
| nsv528228 | Chr.X:151905576 - 151943206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map