Assay Details
Target Gene Details
Entrez Gene ID: | 79608 |
Gene Name: | RIC3 acetylcholine receptor chaperone |
Gene Aliases: |
AYST720, PRO1385 |
Location: |
Chr.11:8092965-8169088 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIC3 |
Target Gene Details
Entrez Gene ID: | 7275 |
Gene Name: | tubby bipartite transcription factor |
Gene Aliases: |
RDOB, rd5 |
Location: |
Chr.11:8019180-8106107 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUB | NM_003320.4 | NP_003311.2 | ||
| NM_177972.2 | NP_813977.1 | |||
| XM_005253109.3 | XP_005253166.1 | |||
| XM_011520344.2 | XP_011518646.1 | |||
| AB209618.1 | 1 | 5840 | BAD92855.1 | |
| AK293528.1 | ||||
| BC075031.2 | AAH75031.1 | |||
| BC075032.2 | AAH75032.1 | |||
| U54644.1 | AAB53494.1 | |||
| U82467.1 | AAB53699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428249 | Chr.11:8047047 - 8194822 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map