Assay Details
Target Gene Details
Entrez Gene ID: | 1735 |
Gene Name: | deiodinase, iodothyronine, type III |
Gene Aliases: |
5DIII, D3, DIOIII, TXDI3 |
Location: |
Chr.14:101561351-101563452 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO3 | NM_001362.3 | 1 | 1092 | NP_001353.4 |
| AK292310.1 | 1 | 948 | ||
| BC017717.1 | 1 | 940 | AAH17717.2 | |
| BX161470.1 | 1 | 1092 | CAD61926.1 | |
| S79854.1 | 1 | 1092 | AAB35616.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047869 | Chr.14:101506888 - 101624595 on Build GRCh38 | Gain |
|
| nsv565882 | Chr.14:101560397 - 101562940 on Build GRCh38 | Loss |
|
| nsv827060 | Chr.14:101559113 - 101566443 on Build GRCh38 | Gain |
|
| nsv952247 | Chr.14:101543464 - 101564963 on Build GRCh38 | Deletion |
|
| nsv565880 | Chr.14:101539186 - 101625954 on Build GRCh38 | Loss |
|
| dgv1978n100 | Chr.14:101504179 - 101603185 on Build GRCh38 | Gain |
|
| esv3635530 | Chr.14:101562410 - 101628188 on Build GRCh38 | Gain |
|
| dgv388n67 | Chr.14:101556368 - 101565620 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map