Assay Details
Target Gene Details
Entrez Gene ID: | 23317 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C13 |
Gene Aliases: |
PARK21, RME8 |
Location: |
Chr.3:132417527-132539032 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC13 | NM_015268.3 | NP_056083.3 | ||
| XM_005247245.4 | XP_005247302.1 | |||
| AB014578.2 | ||||
| AY779857.1 | AAV41096.1 | |||
| BC043583.1 | AAH43583.1 | |||
| BC151246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829728 | Chr.3:132359717 - 132563778 on Build GRCh38 | Gain |
|
| nsv522333 | Chr.3:132431339 - 132441869 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map