Assay Details
Target Gene Details
Entrez Gene ID: | 10494 |
Gene Name: | serine/threonine kinase 25 |
Gene Aliases: |
SOK1, YSK1 |
Location: |
Chr.2:241494707-241509730 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759120 | Chr.2:241480540 - 241562243 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map