Assay Details
Target Gene Details
Entrez Gene ID: | 92689 |
Gene Name: | family with sequence similarity 114 member A1 |
Gene Aliases: |
Noxp20 |
Location: |
Chr.4:38867733-38945744 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM114A1 | NM_138389.2 | NP_612398.2 | ||
| NR_033290.1 | ||||
| XM_005262672.2 | XP_005262729.1 | |||
| XM_006714033.2 | XP_006714096.1 | |||
| XM_017008838.1 | XP_016864327.1 | |||
| AK292831.1 | ||||
| AK296859.1 | ||||
| AK304050.1 | ||||
| BC001096.2 | AAH01096.2 | |||
| BC040452.1 | AAH40452.1 | |||
| BX640862.1 | CAE45923.1 |
More Information
Set Membership: |
|

Genomic Map