Assay Details
Target Gene Details
Entrez Gene ID: | 1141 |
Gene Name: | cholinergic receptor nicotinic beta 2 subunit |
Gene Aliases: |
EFNL3, nAChRB2 |
Location: |
Chr.1:154567781-154580026 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNB2 | NM_000748.2 | 6 | 1724 | NP_000739.1 |
| XM_017000180.1 | 6 | 1570 | XP_016855669.1 | |
| AK313470.1 | ||||
| BC075040.2 | 6 | 1489 | AAH75040.1 | |
| BC075041.2 | 6 | 1489 | AAH75041.1 | |
| U62437.1 | 6 | 1724 | AAB40115.1 | |
| X53179.1 | 6 | 1626 | CAA37320.1 | |
| Y08415.1 | CAA69692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161683 | Chr.1:154575785 - 154587122 on Build GRCh38 | Duplication |
|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map