Assay Details
Target Gene Details
Entrez Gene ID: | 101927583 |
Gene Name: | uncharacterized LOC101927583 |
Gene Aliases: |
- |
Location: |
Chr.12:57618405-57621903 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927583 | NR_120450.1 | 2 | 1794 |
Target Gene Details
Entrez Gene ID: | 65012 |
Gene Name: | solute carrier family 26 member 10 |
Gene Aliases: |
- |
Location: |
Chr.12:57619910-57626151 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A10 | NM_133489.2 | 1 | 48 | NP_597996.2 |
| AF331523.1 | 1 | 48 | AAM92901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map