Assay Details
Target Gene Details
Entrez Gene ID: | 79176 |
Gene Name: | F-box and leucine rich repeat protein 15 |
Gene Aliases: |
FBXO37, Fbl15, JET, PSD |
Location: |
Chr.10:102419222-102423138 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL15 | NM_024326.3 | 1 | 589 | NP_077302.3 |
| XM_005270149.2 | XP_005270206.1 | |||
| XM_005270150.4 | 1 | 477 | XP_005270207.1 | |
| XM_005270151.3 | XP_005270208.1 | |||
| XM_017016631.1 | 1 | 122 | XP_016872120.1 | |
| AK122684.1 | 2 | 685 | ||
| BC036120.1 | 1 | 571 | AAH36120.1 | |
| BC130566.1 | ||||
| BI914195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159761 | Chr.10:102372778 - 102430809 on Build GRCh38 | Deletion |
|
| nsv951006 | Chr.10:102394644 - 102425743 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115842294] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map