Assay Details
Target Gene Details
Entrez Gene ID: | 8658 |
Gene Name: | tankyrase |
Gene Aliases: |
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5 |
Location: |
Chr.8:9555229-9782346 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS | NM_003747.2 | 2 | 763 | NP_003738.2 |
| XM_006716263.3 | 2 | 1469 | XP_006716326.1 | |
| XM_011543845.2 | 2 | 1469 | XP_011542147.1 | |
| XM_011543846.2 | 2 | 1469 | XP_011542148.1 | |
| XM_011543847.2 | 2 | 1468 | XP_011542149.1 | |
| AF082556.1 | 2 | 763 | AAC79841.1 | |
| AF082557.1 | 1 | 59 | AAC79842.1 | |
| AK301798.1 | 2 | 684 | ||
| AK302031.1 | 2 | 784 | ||
| AK302534.1 | 2 | 437 | ||
| BC098394.1 | 2 | 786 | AAH98394.1 |
More Information
Set Membership: |
|

Genomic Map