Assay Details
Target Gene Details
Entrez Gene ID: | 347169 |
Gene Name: | olfactory receptor family 1 subfamily B member 1 (gene/pseudogene) |
Gene Aliases: |
OR9-26, OR9-B |
Location: |
Chr.9:122578405-122657626 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1B1 |
Target Gene Details
Entrez Gene ID: | 158131 |
Gene Name: | olfactory receptor family 1 subfamily Q member 1 |
Gene Aliases: |
HSTPCR106, OR1Q2, OR1Q3, OR9-25, OR9-A, OST226, OST226OR9-A, TPCR106 |
Location: |
Chr.9:122614738-122615682 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1Q1 | NM_012364.1 | 1 | 378 | NP_036496.1 |
| BC140713.1 | 1 | 447 | ||
| X89667.1 | CAA61814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054800 | Chr.9:122555206 - 122666496 on Build GRCh38 | Loss |
|
| nsv1039785 | Chr.9:122544123 - 122644710 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map