Assay Details
Target Gene Details
Entrez Gene ID: | 374868 |
Gene Name: | ATPase phospholipid transporting 9B (putative) |
Gene Aliases: |
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1 |
Location: |
Chr.18:79069275-79378391 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP9B | NM_001306085.1 | 2 | 380 | NP_001293014.1 |
| NM_198531.4 | 2 | 380 | NP_940933.3 | |
| XM_011525963.2 | 3 | 546 | XP_011524265.1 | |
| XM_011525964.2 | 2 | 261 | XP_011524266.1 | |
| XM_011525965.2 | 3 | 542 | XP_011524267.1 | |
| XM_011525966.2 | 2 | 497 | XP_011524268.1 | |
| XM_017025726.1 | 3 | 546 | XP_016881215.1 | |
| XM_017025727.1 | 3 | 546 | XP_016881216.1 | |
| XM_017025728.1 | 3 | 546 | XP_016881217.1 | |
| XM_017025729.1 | 3 | 546 | XP_016881218.1 | |
| XM_017025730.1 | 3 | 546 | XP_016881219.1 | |
| XM_017025731.1 | 3 | 546 | XP_016881220.1 | |
| XM_017025732.1 | 3 | 546 | XP_016881221.1 | |
| XM_017025733.1 | 3 | 546 | XP_016881222.1 | |
| XM_017025734.1 | 3 | 546 | XP_016881223.1 | |
| XM_017025735.1 | 3 | 546 | XP_016881224.1 | |
| XM_017025736.1 | 3 | 546 | XP_016881225.1 | |
| XM_017025737.1 | 3 | 546 | XP_016881226.1 | |
| XM_017025742.1 | 3 | 546 | XP_016881231.1 | |
| AB373110.1 | 1 | 88 | ||
| AK093704.1 | ||||
| AK097757.1 | 2 | 497 | ||
| AK295981.1 | 2 | 247 | ||
| AK308291.1 | 2 | 262 | ||
| BC053561.1 | 2 | 165 | AAH53561.1 | |
| BC125219.1 | 2 | 258 | ||
| BC125220.1 | 2 | 258 | ||
| HY028472.1 | 2 | 380 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065064 | Chr.18:79091898 - 79405383 on Build GRCh38 | Gain |
|
| nsv577794 | Chr.18:78951366 - 79472709 on Build GRCh38 | Gain |
|
| esv3643288 | Chr.18:79087927 - 79251969 on Build GRCh38 | Loss |
|
| nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain |
|
| esv3643290 | Chr.18:79088460 - 79140822 on Build GRCh38 | Loss |
|
| esv3643283 | Chr.18:79003881 - 79112789 on Build GRCh38 | Loss |
|
| esv2717497 | Chr.18:78170182 - 79635788 on Build GRCh38 | Deletion |
|
| nsv1141945 | Chr.18:78170394 - 79635820 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map