Assay Details
Target Gene Details
Entrez Gene ID: | 221074 |
Gene Name: | solute carrier family 39 member 12 |
Gene Aliases: |
LZT-Hs8, ZIP-12, bA570F3.1 |
Location: |
Chr.10:17951839-18043292 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A12 | NM_001145195.1 | 2 | 310 | NP_001138667.1 |
| NM_001282733.1 | 2 | 310 | NP_001269662.1 | |
| NM_001282734.1 | NP_001269663.1 | |||
| NM_152725.3 | 2 | 310 | NP_689938.2 | |
| AK055061.1 | 2 | 258 | BAB70848.1 | |
| AK295294.1 | ||||
| AK303735.1 | ||||
| BC035118.1 | 2 | 242 | AAH35118.1 | |
| BC047635.1 | 2 | 225 | AAH47635.1 | |
| BC065917.1 | 1 | 54 | AAH65917.1 | |
| BC094700.1 | 2 | 186 | AAH94700.1 | |
| BC117323.1 | 1 | 60 | ||
| BC143551.1 | 1 | 59 | ||
| DB469365.1 | 2 | 312 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039655 | Chr.10:17941330 - 18060025 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map