Assay Details
Target Gene Details
Entrez Gene ID: | 84364 |
Gene Name: | ADP ribosylation factor GTPase activating protein 2 |
Gene Aliases: |
IRZ, NBLA10535, ZFP289, ZNF289 |
Location: |
Chr.11:47164298-47177503 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFGAP2 | NM_001242832.1 | NP_001229761.1 | ||
| NM_032389.4 | NP_115765.2 | |||
| XM_005253166.1 | XP_005253223.1 | |||
| XM_005253167.1 | XP_005253224.1 | |||
| XM_005253168.2 | XP_005253225.1 | |||
| XM_006718346.1 | XP_006718409.1 | |||
| XM_011520405.1 | XP_011518707.1 | |||
| XM_017018413.1 | XP_016873902.1 | |||
| XM_017018414.1 | XP_016873903.1 | |||
| XM_017018415.1 | XP_016873904.1 | |||
| AB073358.1 | BAE45716.1 | |||
| AK027482.1 | BAB55144.1 | |||
| AK098324.1 | ||||
| AK301785.1 | ||||
| AK304541.1 | ||||
| AK315992.1 | ||||
| AL834337.1 | CAD39004.1 | |||
| BC030148.2 | AAH30148.1 | |||
| BM827445.1 |
More Information
Set Membership: |
|

Genomic Map