Assay Details
Target Gene Details
Entrez Gene ID: | 3460 |
Gene Name: | interferon gamma receptor 2 (interferon gamma transducer 1) |
Gene Aliases: |
AF-1, IFGR2, IFNGT1, IMD28 |
Location: |
Chr.21:33402896-33437521 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNGR2 | NM_005534.3 | 7 | 1718 | NP_005525.2 |
| XM_005260969.2 | 8 | 1234 | XP_005261026.1 | |
| XM_011529553.1 | 8 | 1160 | XP_011527855.1 | |
| XM_011529554.2 | 7 | 1594 | XP_011527856.1 | |
| AI824138.1 | 1 | 517 | ||
| AK292246.1 | 7 | 1166 | ||
| BC003624.2 | 7 | 1186 | AAH03624.1 | |
| BQ008968.1 | 1 | 521 | ||
| U05875.1 | 7 | 1718 | AAA16955.1 | |
| U05877.1 | 7 | 1183 | AAA16956.1 |
Target Gene Details
Entrez Gene ID: | 757 |
Gene Name: | transmembrane protein 50B |
Gene Aliases: |
C21orf4, HCVP7TP3 |
Location: |
Chr.21:33432486-33480009 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM50B | NR_040016.1 | |||
| XM_011529746.1 | XP_011528048.1 | |||
| CB306801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834087 | Chr.21:33325890 - 33469002 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map