Assay Details
Target Gene Details
Entrez Gene ID: | 94086 |
Gene Name: | heat shock protein family B (small) member 9 |
Gene Aliases: |
CT51 |
Location: |
Chr.17:42122738-42123353 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB9 | NM_033194.2 | 1 | 236 | NP_149971.1 |
| AK093488.1 | 1 | 1544 | ||
| AK130947.1 | 1 | 144 | ||
| BC093991.1 | 1 | 157 | AAH93991.1 | |
| BC093993.1 | 1 | 157 | AAH93993.1 | |
| BX280091.1 | 1 | 236 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953898 | Chr.17:42113083 - 42131582 on Build GRCh38 | Deletion |
|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map