Assay Details
Target Gene Details
Entrez Gene ID: | 29888 |
Gene Name: | striatin 4 |
Gene Aliases: |
PPP2R6C, ZIN, zinedin |
Location: |
Chr.19:46719509-46746475 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRN4 | NM_001039877.1 | 5 | 612 | NP_001034966.1 |
| NM_013403.2 | 5 | 612 | NP_037535.2 | |
| XM_006723171.2 | 6 | 669 | XP_006723234.1 | |
| XM_006723172.2 | 6 | 669 | XP_006723235.1 | |
| XM_006723174.2 | 5 | 419 | XP_006723237.1 | |
| XM_011526878.1 | 6 | 669 | XP_011525180.1 | |
| XM_017026716.1 | 6 | 398 | XP_016882205.1 | |
| XM_017026717.1 | 5 | 374 | XP_016882206.1 | |
| XM_017026719.1 | 5 | 624 | XP_016882208.1 | |
| AB209003.1 | 5 | 564 | BAD92240.1 | |
| AF212940.1 | 5 | 579 | AAF29527.1 | |
| AK297637.1 | 6 | 591 | ||
| AK298804.1 | 6 | 519 | ||
| BC034604.1 | 5 | 589 | AAH34604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map