Assay Details
Target Gene Details
Entrez Gene ID: | 140459 |
Gene Name: | ankyrin repeat and SOCS box containing 6 |
Gene Aliases: |
- |
Location: |
Chr.9:129634604-129642169 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB6 | NM_001202403.1 | 5 | 3097 | NP_001189332.1 |
| NM_017873.3 | 6 | 3184 | NP_060343.1 | |
| NM_177999.2 | 5 | 3075 | NP_821066.1 | |
| AK093539.1 | 1 | 732 | ||
| AL390135.1 | 1 | 1457 | ||
| BX648923.1 | 5 | 3705 | CAH10563.1 | |
| CR627462.1 | 1 | 1457 |
Target Gene Details
Entrez Gene ID: | 28989 |
Gene Name: | N-terminal Xaa-Pro-Lys N-methyltransferase 1 |
Gene Aliases: |
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A |
Location: |
Chr.9:129608884-129636742 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTMT1 | NM_001286796.1 | 4 | 1599 | NP_001273725.1 |
| NM_001286797.1 | 4 | 1412 | NP_001273726.1 | |
| NM_001286798.1 | 4 | 1473 | NP_001273727.1 | |
| NM_001286799.1 | 4 | 1402 | NP_001273728.1 | |
| NM_001286800.1 | 4 | 1362 | NP_001273729.1 | |
| NM_001286801.1 | 4 | 1388 | NP_001273730.1 | |
| NM_001286802.1 | 3 | 1257 | NP_001273731.1 | |
| NM_001286803.1 | 3 | 1231 | NP_001273732.1 | |
| NM_014064.3 | 4 | 1447 | NP_054783.2 | |
| NR_104596.1 | 4 | 1298 | ||
| XM_005251939.3 | 4 | 1388 | XP_005251996.1 | |
| XM_017014642.1 | 4 | 1305 | XP_016870131.1 | |
| AK298840.1 | 4 | 1352 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469901 | Chr.9:129597897 - 129681820 on Build GRCh38 | Loss |
|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv951784 | Chr.9:129625222 - 129655421 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map