Assay Details
Target Gene Details
Entrez Gene ID: | 200010 |
Gene Name: | solute carrier family 5 member 9 |
Gene Aliases: |
SGLT4 |
Location: |
Chr.1:48221113-48248644 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A9 | NM_001011547.2 | NP_001011547.2 | ||
| NM_001135181.1 | NP_001128653.1 | |||
| XM_011540924.2 | XP_011539226.1 | |||
| XM_011540925.2 | XP_011539227.1 | |||
| XM_011540926.2 | XP_011539228.1 | |||
| XM_011540927.1 | XP_011539229.1 | |||
| XM_011540928.1 | XP_011539230.1 | |||
| XM_011540929.2 | XP_011539231.1 | |||
| XM_017000558.1 | XP_016856047.1 | |||
| AK131200.1 | ||||
| AK296133.1 | ||||
| BC104857.1 | AAI04858.1 | |||
| BC104863.1 | AAI04864.1 | |||
| BC143570.1 | ||||
| BC143572.1 | ||||
| BX648549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578249 | Chr.1:48202315 - 48332128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map