Assay Details
Target Gene Details
Entrez Gene ID: | 253943 |
Gene Name: | YTH N6-methyladenosine RNA binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.8:63168553-63212788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YTHDF3 | NM_001277813.1 | 1 | 90 | NP_001264742.1 |
| NM_001277815.1 | 1 | 90 | NP_001264744.1 | |
| NM_001277816.1 | 1 | 90 | NP_001264745.1 | |
| NM_001277817.1 | NP_001264746.1 | |||
| NM_152758.5 | 1 | 90 | NP_689971.4 | |
| NR_102434.1 | 1 | 90 | ||
| XM_011517509.2 | XP_011515811.1 | |||
| XM_011517510.2 | XP_011515812.1 | |||
| AK056219.1 | BAB71122.1 | |||
| AK127574.1 | ||||
| AL831816.1 | CAD38530.2 | |||
| AL832005.1 | CAH56224.1 | |||
| AL832335.1 | CAH56223.1 | |||
| AW673093.1 | ||||
| BC052970.1 | 1 | 39 | AAH52970.1 | |
| BP223018.1 | 1 | 90 | ||
| BP354748.1 | ||||
| BX537996.2 | CAH56480.1 | |||
| CN335163.1 | ||||
| DC419051.1 |
More Information
Set Membership: |
|

Genomic Map