Assay Details
Target Gene Details
Entrez Gene ID: | 119032 |
Gene Name: | BLOC-1 related complex subunit 7 |
Gene Aliases: |
C10orf32 |
Location: |
Chr.10:102854210-102864961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS7 | NM_001136200.1 | NP_001129672.1 | ||
| NM_144591.3 | NP_653192.2 | |||
| AK098071.1 | ||||
| AK311742.1 | ||||
| AL832477.1 | CAI46115.1 | |||
| BC015994.1 | AAH15994.1 | |||
| DA006534.1 |
Target Gene Details
Entrez Gene ID: | 100528007 |
Gene Name: | BORCS7-ASMT readthrough (NMD candidate) |
Gene Aliases: |
C10orf32-AS3MT, C10orf32-ASMT |
Location: |
Chr.10:102854210-102901899 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS7-ASMT | NR_037644.1 |
More Information
Set Membership: |
|

Genomic Map