Assay Details
Target Gene Details
Entrez Gene ID: | 29086 |
Gene Name: | BRISC and BRCA1 A complex member 1 |
Gene Aliases: |
C19orf62, HSPC142, MERIT40, NBA1 |
Location: |
Chr.19:17267376-17279353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BABAM1 | NM_001033549.2 | NP_001028721.1 | ||
| NM_001288756.1 | NP_001275685.1 | |||
| NM_001288757.1 | NP_001275686.1 | |||
| NM_014173.3 | NP_054892.2 | |||
| AF161491.1 | AAF29106.1 | |||
| AK000578.1 | BAA91268.1 | |||
| AK025970.1 | ||||
| AK225047.1 | ||||
| AK301193.1 | ||||
| AK308590.1 | ||||
| AL136692.1 | CAB66627.1 | |||
| BC000788.2 | AAH00788.1 | |||
| BC006244.1 | AAH06244.1 | |||
| BC091491.1 | AAH91491.1 | |||
| CR533526.1 | CAG38557.1 | |||
| CX866662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578740 | Chr.19:17259381 - 17285740 on Build GRCh38 | Loss |
|
| nsv515965 | Chr.19:17273458 - 17309480 on Build GRCh38 | Loss |
|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map