Assay Details
Target Gene Details
Entrez Gene ID: | 9232 |
Gene Name: | pituitary tumor-transforming 1 |
Gene Aliases: |
EAP1, HPTTG, PTTG, TUTR1 |
Location: |
Chr.5:160421807-160428744 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTTG1 | NM_001282382.1 | NP_001269311.1 | ||
| NM_001282383.1 | NP_001269312.1 | |||
| NM_004219.3 | NP_004210.1 | |||
| AF062649.1 | AAC69752.1 | |||
| AF075242.1 | AAD19335.1 | |||
| AF095287.1 | AAC64409.1 | |||
| AJ223953.1 | CAA11683.1 | |||
| BC101834.1 | AAI01835.1 | |||
| BC112071.1 | AAI12072.1 | |||
| BM990391.1 | ||||
| BU182149.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140989544] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map